Learn More
Thermo Scientific™ Pierce™ Premium Grade TCEP-HCl
Optimize applications where product integrity and risk minimization are paramount with our highest quality formulation of this disulfide reducing agent.
Brand: Thermo Scientific™ PG82089
Code : NEW
Additional Details : Weight : 0.01000kg Transport : UN number : 3261 Chem class : 8 Pack group : II
Description
Optimize applications where product integrity and risk minimization are paramount with the specially characterized Thermo Scientific™ Pierce™ Premium Grade TCEP-HCl, our highest quality formulation of this disulfide reducing agent.
Thermo Scientific Pierce Premium Grade TCEP-HCl is our highest quality formulation of this disulfide reducing agent, specially characterized for applications where product integrity and risk minimization are paramount.
Compared to the standard grade product, Premium Grade SPDP provides more clearly defined quality and product support by including (a) increased analytical testing and product characterization, (b) greater batch-specific information and quality assurance review, (c) extensive lot sample retention and (d) change control notification. TCEP-HCl is pure, crystalline Tris(2-carboxyethyl)phosphine hydrochloride, a thiol-free compound that is highly effective at reducing protein and peptide disulfide bonds, as well as being odorless and stable. TCEP selectively and completely reduces even the most stable water-soluble alkyl disulfides over a wide pH range.
Highlights:- TCEP – a thiol-free, odorless yet potent reducing agent for cleaving disulfide bonds
- High quality – identity and purity confirmed by several tests, including quantitative NMR
- Product integrity – enhanced level of testing and characterization compared to standard grade
- Lot retention – ample supply of past lots retained to ensure future process testing
- Change management – Change Control Notification (CCN) service
- Consistent manufacture – batch-specific manufacturing documentation review
- Odorless – Unlike DTT or BME, TCEP is odor-free, so reductions can be carried out conveniently on the bench top.
- Stable in air – The inherent stability of the TCEP moiety eliminates the need for any special precautions to avoid oxidation when handling, using or storing TCEP.
- Efficient – For most applications, 5 to 50mM TCEP provides sufficient molar excess to effectively reduce peptide or protein disulfide bonds within a few minutes at room temperature.
- Compatible – With TCEP, removal of the reducing agent is not necessary prior to most applications, (e.g. histidine-tagged protein purification, maleimide conjugations).
Specifications
Upon receipt store product at room temperature in sealed container to prevent oxidation. Product is shipped at ambient temperature. | |
Solid | |
Disulfide Reducing Reagent |
Pierce™ | |
10 g |
Safety and Handling
- Pierce Premium Grade TCEP-HCl
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H290-May be corrosive to metals.
- H318-Causes serious eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : 3,3',3''-phosphinidynetris-propanoic acid hydrochloride
For Research Use Only. Not for use in diagnostic procedures.