Learn More
Thermo Scientific™ Pierce™ Phosphoprotein Enrichment Kit
Purify (enrich) phosphorylated proteins from mammalian cell and tissue lysates using phospho-specific IMAC affinity resin cartidges or kit.
Brand: Thermo Scientific™ 90003
554.80 GBP valid until 2024-03-29
Use promo code "21653" to get your promotional price.
Code : PN
Additional Details : Weight : 0.10000kg
Description
Perform efficient enrichment of phosphorylated proteins derived from mammalian cells and tissues with Thermo Scientific™ Pierce Phosphoprotein Enrichment Kit.
The kit is an effective system for isolating the phosphoprotein fraction of mammalian cell and tissue lysates. The included lysis-binding buffer and proprietary affinity resin produce superior yields with negligible non-specific binding. The kit uses a convenient spin column format, and the resulting phosphorylated protein sample is compatible with downstream applications, including Western blotting, mass spectrometry (MS), 2D-PAGE, and protein arrays. The affinity resin is also available as pre-packed chromatography cartridges for use with automated liquid chromatography (LC) systems.Highlights:
Specific – low nonspecific protein contamination from complex biological samples, such as cell culture lysate and mouse tissue extract
Fast – easy spin format enables enrichment of phosphorylated proteins in less than 2 hours
Superior yield – achieves higher yields than other commercially available phosphoprotein isolation kits
Convenient format – complete kit offering pre-dispensed spin columns, buffers, reagents and ultrafiltration columns for enrichment
Compatible – downstream applications include mass spectrometry, Western blotting and 2D-PAGE
Specifications
Upon receipt store at 4°C. | |
10 Column Kit | |
Phosphoprotein Enrichment Kit | |
10 Reactions | |
Affinity, Agarose Resin |
Pierce™ | |
Phosphoprotein | |
Proprietary Ligand | |
Spin Column |
For Research Use Only. Not for use in diagnostic procedures.