Learn More
Thermo Scientific™ Pierce™ MS Grade Trypsin Protease
Ensure maximum activity and stability in proteomic applications with this chemically modified, highly purified porcine trypsin.
£47.56 - £397.00
Products 4
Product Code Brand Description Quantity Price Quantity & Availability  
Product Code Brand Description Quantity Price Quantity & Availability  
13454189 View Documents Thermo Scientific™
90058
Pierce Trypsin Protease, MS Grade 5 x 100µg £205.00
Pack of 5 View alternative products
Save Up to 
»
EU Stock 66
Add to basket
13464189 View Documents Thermo Scientific™
90057
Pierce Trypsin Protease, MS Grade 5 x 20µg £47.56
Pack of 5 View alternative products
Save Up to 
»
EU Stock 89
Add to basket
13474189 View Documents Thermo Scientific™
90059
Pierce Trypsin Protease, MS Grade 1mg £397.00
1mg View alternative products
Save Up to 
»
EU Stock 22
Add to basket
15255753 View Documents Thermo Scientific™
90305
Pierce Trypsin Protease, MS Grade, Frozen Liquid 1 x 100μg £58.75
100µg View alternative products
Save Up to 
»
EU Stock 70
Add to basket
Description
Description
Pierce Trypsin Protease, MS Grade, is a serine protease derived from porcine pancreatic extracts. The enzyme is TPCK-treated to eliminate chymotryptic activity and methylated to improve stability during protein digestion. This mass spectrometry-grade, modified trypsin is then repurified, lyophilized in glass vials and packaged in convenient 5 x 20μg, 5 x 100μg, and 1mg fill sizes.
Highlights:
  • Exceptional selectivity – cleaves at the carboxyl side of lysine and arginine residues with greater than 95% specificity
  • High purity – no detectable chymotrypsin activity
  • Enhanced stability – modified and lyophilized for reduced autolytic activity
  • Economical – available in larger fill sizes that are more cost effective
Recommended for:
  • In-gel digestion of proteins from 1–D or 2–D gels
  • In-solution tryptic digestion of proteins