Learn More
Pierce™ Methyl-PEG-Amine Compounds
MA(PEG)n: methyl- and amine-capped polyethylene glycol compounds containing 4 to 24 PEG units; use for surface-modification and molecule-pegylation applications.
Brand:  Thermo Scientific Pierce 26116
Code : PN
Additional Details : CAS Number : 175205-42-2 Weight : 0.80000kg
 View more versions of this product
Product Code. 11821415
  • £247.76 / 100mg
Estimated Shipment
21-03-2018
Add to basket
Description
Description
MA(PEG)n compounds have the form Methyl-PEGn-Amine and are ideal for derivatizing surfaces and molecules with discrete numbers (n = 4, 8, 12, 24) of ethylene glycol groups. Use any amine-reactive chemistry (e.g., EDC or NHS ester) to PEGylate surfaces, proteins, or other molecules with a precise number of hydrophilic, nonimmunogenic PEG units.
Highlights:
  • Pure compounds of specific size ensure complete control of PEGylation
  • Straight-chain compounds with simple terminal primary amine
  • Adaptable to a variety of modification procedures
Recommended for:
  • PEGylate carboxylate or amine surfaces Add inert mass to proteins, immunogens, drug compounds and probes Improve solubility (decrease aggregation) of proteins or peptides without affecting function Protect proteins from proteolysis
Specifications
Specifications
Methyl
No
24-unit ethyleneglycol functionalized with methyl and amine ends; waxy solid
Yes
4mL of typical 1X (25mM) reaction solution
Carboxyl (via EDC)
Pierce MA(PEG)24
1088.32
100mg
86.1Å
Videos