Learn More
Thermo Scientific™ Pierce™ MALDI Matrices
Highly purified, recrystallized CHCA, DHB and SA reagents in single-use tubes for mass spectrometry by matrix-assisted laser desorption ionization (MALDI).
£163.00 - £167.00
Specifications
Quantity 24 x 1mg
Sufficient For Analysis of up to 96 samples with each tube after dissolving its contents in 100µL of solvent
View More Specs
Products 4
Product Code Brand Description Price Quantity & Availability  
Product Code Brand Description Price Quantity & Availability  
10424375 View Documents Thermo Scientific™
90031
Pierce CHCA MALDI Matrix £164.00
Pack of 3 View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 06-01-2020
10434375 View Documents Thermo Scientific™
90032
Pierce SA MALDI Matrices £164.00
1 set View alternative products
Save Up to 
»
Estimated Shipment date
from Supplier 06-01-2020
10046153 View Documents Thermo Scientific™
90033
Pierce DHB MALDI Matrices £163.00
Pack of 5 View alternative products
Save Up to 
»
EU Stock 3 Please to purchase this item. Need a web account? Register with us today!
11865834 View Documents Thermo Scientific™
90035
Pierce MALDI Matrix Sample Pack £167.00
Pack of 8 View alternative products
Save Up to 
»
EU Stock 3 Please to purchase this item. Need a web account? Register with us today!
Description
Description
The MALDI method for MS peptide analysis is now easier to perform with single-use, highly pure MALDI matrices that are exceptionally convenient to use. Matrix assisted laser desorption ionization (MALDI) matrices produce the cleanest mass spectrometric (MS) spectra when they are recrystallized and prepared daily, which is inconvenient and typically results in significant waste. The packaging of purified, recrystallized alpha-cyano-4-hydroxy-cinnamic acid (CHCA), sinapinic acid (SA) and 2,5-dihydroxybenzoic acid (DHB) MALDI matrices makes preparation of high-quality MALDI reagents easy.
Highlights:
  • Highly purified, recrystallized reagent
  • Packaged in convenient single-use tubes sufficient for analyzing up to 96 samples to save time and reduce waste
  • Provides accurate quantity of matrix every time
Specifications
Specifications
24 x 1mg
Analysis of up to 96 samples with each tube after dissolving its contents in 100µL of solvent