Learn More
Thermo Scientific™ MALDI Matrix Sample Pack, Single-Use
Highly purified, recrystallized CHCA, DHB and SA reagents in single-use tubes for mass spectrometry by matrix-assisted laser desorption ionization (MALDI).
Brand: Thermo Scientific™ 90035
Code : PN
Additional Details : CAS Number : 1500-93-2 Weight : 0.80000kg
Description
Thermo Scientific™ MALDI Matrices are highly purified, recrystallized reagents packaged in convenient single-use tubes sufficient for analyzing up to 96 samples by matrix assisted laser desorption ionization.
The MALDI method for MS peptide analysis is now easier to perform with single-use, highly pure MALDI matrices that are exceptionally convenient to use. Matrix assisted laser desorption ionization (MALDI) matrices produce the cleanest mass spectrometric (MS) spectra when they are recrystallized and prepared daily, which is inconvenient and typically results in significant waste. The packaging of purified, recrystallized alpha-cyano-4-hydroxy-cinnamic acid (CHCA), sinapinic acid (SA) and 2,5-dihydroxybenzoic acid (DHB) MALDI matrices makes preparation of high-quality MALDI reagents easy.Highlights:
- Highly purified, recrystallized reagent
- Packaged in convenient single-use tubes sufficient for analyzing up to 96 samples to save time and reduce waste
- Provides accurate quantity of matrix every time
Specifications
Pierce MALDI Matrix Sample Pack | |
MALDI Matrix | |
Small Molecules | |
Mass Spectrometry |
Mass Spectrometer | |
Upon receipt store product at 4°C. | |
Small Molecules | |
Calibration |
For Research Use Only. Not for use in diagnostic procedures.