Learn More
Thermo Scientific™ Pierce™ Krypton™ Fluorescent Protein Stain
Stain proteins in polyacrylamide gels or transfer membranes for fluorescent detection with popular fluorescence imagers.
Brand:  Thermo Scientific™ 46628
Code : PN
Additional Details : Weight : 0.02000kg
 View more versions of this product
Product Code. 10332044
  • £44.09 / 20mL
EU Stock 2
Add to basket
Description
Description
This fluorescent protein gel stain is for bright yellow-orange fluorescent detection of proteins separated by 1D or 2D polyacrylamide gel electrophoresis (SDS-PAGE). Krypton Stain uses a protein-binding dye with fluorescence properties (Ex/Em = 520/580nm) suitable for visualization with a variety of standard laser-based and filter-based fluorescence imagers and documentation systems. Proteins are detectable at concentrations greater than 0.25ng per band in a typical mini-gel lane (greater than 2ng for the rapid staining protocol). Krypton Stain provides high signal intensity, exhibits minimal protein-to-protein variation, has an excellent quantitative dynamic range and is compatible with subsequent destaining and analysis by mass spectrometry.
Highlights:
  • Intense fluorescent stain – excitation and emission maxima of 520nm and 580nm, respectively (compatible with 532nm excitation light sources and 580 to 600nm emission filters)
  • Instrument-compatible – effective with popular laser-based and filter-based fluorescence imagers
  • Broad dynamic range – linear quantitative range spans three to four orders of magnitude
  • Sensitive – detect as little as 0.25ng protein with the standard protocol (approx. 2.5 hours); at least as sensitive as fluorescent stains from other suppliers
  • Fast – standard (2.5 hours) and rapid (30 minutes) protocols are significantly faster than procedures with other popular fluorescent stains
  • Versatile – compatible with proteomics workflows involving downstream mass spectrometry
  • Compact – supplied as a 10X solution to save storage space; just dilute with water before use
Specifications
Specifications
Krypton Fluorescent Protein Stain
Proprietary solution containing fluorescent protein-binding dye
20mL
4 mini gels (SDS-PAGE)
Videos