Learn More
Thermo Scientific™ Pierce™ High Binding NeutrAvidin™ Coated Plates
Attain higher biotin-binding efficiency with these pre-blocked, ready-to-use high-capacity NeutrAvidin protein-coated microplates.
Brand:  Thermo Scientific™ 15512
Code : PN
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10177573
  • £284.08 / Pack of 5
Estimated Shipment
06-02-2018
Add to basket
Description
Description
Biotin-binding plates are ideal for applications when direct adsorption to polystyrene denatures antibodies or the target molecule. Thermo Scientific NeutrAvidin™ Protein is avidin that has been deglycosylated to prevent nonspecific lectin-binding. NeutrAvidin Protein has a near-neutral pI (6.3), resulting in lower nonspecific interactions in certain applications compared with streptavidin. NeutrAvidin Protein has an additional advantage of lacking the bacterial RYD sequence found on streptavidin, which eliminates nonspecific binding due to the RGD binding domain of adhesion receptors present in a variety of cells. Pierce NeutrAvidin High Binding Capacity (HBC) Coated Plates are available in clear, white and black plates for use with colorimetric, chemiluminescent and fluorescent detection methods, respectively.
 Highlights:
  • Exclusive plate-coating technology – approx. twice the capacity of typical biotin-binding plates
  • Robust and sensitive – NeutrAvidin Protein yields less nonspecific binding for improved signal-to-noise ratios
  • Broad dynamic range – extends the quantitative range so there is no need for dilutions
  • Save time – pre-blocked plates (two types available) reduces the number of assay steps
  • Flexible assay formats – choose 96- and 384-well formats and in different colors
  • NeutrAvidin Protein – performance equals or exceeds streptavidin but costs less
Specifications
Specifications
Biotin
White, 384-well plates, coated with 50uL of NeutrAvidin Protein and blocked with 100uL of SuperBlock Blocking Buffer
Binding approx. 35pmol biotin per well
Pierce NeutrAvidin Coated High Capacity Plates, White, 384-Well
5 plates
Videos
Documentation
Technical Documents