missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Heptafluorobutyric Acid (HFBA)

Bottles and ampules of 99.5%-pure HFBA (MW 214.04, CAS 375-22-4), valid for use as an ion-pairing agent in reverse-phase peptide separations (HPLC).

Brand:  Thermo Scientific™ TS-25003

Code : NEW

Additional Details : Weight : 0.70000kg

 View more versions of this product

Product Code. 10714767

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »




  • Typical purity is 99.7% by GC; <0.1% water
  • Sequencing reagent for classical and automated Edman degradation of peptides and proteins
  • Density: 1.645
  • B.P. 120°C
  • Packaged under nitrogen in amber glass ampules or bottles
  • Clear, colorless liquid

Ordering Information:

X in the ordering table indicates that hazardous shipping charges apply.



Corrosive, Irritant
Heptafluorobutyric acid
Clear, colorless liquid of HFBA (at least 99.5% pure; zero sulfate; low water; low absorbance)
Room Temperature
98 to 100%
Pierce Heptafluorobutyric Acid (HFBA)

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.