Learn More
Thermo Scientific™ Pierce™ GTPase Enrichment Kit and Probes
A desthiobiotin-GTP probe kit to label, affinity purify and measure the active site of GTPases and the GTPase subunits of G-protein coupled receptors.
Brand:  Thermo Scientific™ 88314
Code : PN
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11842134
  • £455.69 / Each
Estimated Shipment
15-06-2018
Add to basket
Description
Description
ActivX GTP Probes feature a desthiobiotin (biotin analog) tag that facilitates selective enrichment and recovery of labeled GTPases allowing identification and profiling of target enzyme classes across samples or to assess the specificity and affinity of enzyme inhibitors.
Highlights:
  • Specific – label only the conserved active-site lysines of nucleotide-binding proteins
  • Compatible – use for in vitro labeling of GTPase enzymes derived from cells or tissues.
  • Flexible – use with Western blot or mass spectrometry (MS) workflows
Recommended for:
Broad enrichment of GTP-binding proteins from tissues, cells and subcellular proteomes; Enrichment of enzymes based on function; Profiling of dozens to hundreds of inhibitor targets.
Specifications
Specifications
Pierce GTPase Enrichment Kit with GTP Probe
Zeba Spin Desalting Columns, 7K MWCO, 8 x 5mL columns, Urea, 12g, Reaction Buffer, 125mL, Pierce IP Lysis Buffer, 100mL, High Capacity Streptavidin Agarose Resin (50% slurry), 1mL, Halt Protease/Phosphatase Inhibitor Cocktail (100X), 1mL, ActivX Desthiobiotin-GTP Probe, 16 x 12.9ug, MgCl2, 500┬ÁL
16 pull-down assays
Multi-component kit with Desthiobiotin-GTP Probe
16 rxn kit
Agarose
Videos