Learn More
Thermo Scientific™ Pierce™ GST (Glutathione S-Transferase)
Pure, lyophilized glutathione S-transferase (GST) protein from Schistosoma japonicum for use in recombinant fusion protein research methods and assays.
Brand:  Thermo Scientific™ 20239
Code : PN
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11826744
  • £843.71 / Pack of 10
Estimated Shipment
08-02-2018
Add to basket
Description
Description
This pure GST from Schistosoma japonicum is useful in a variety of applications involving GST-tagged fusion proteins and glutathione affinity purification. The expression of foreign proteins in E. coli using a glutathione S-transferase (GST) fusion system is a method of choice for studying the function of particular proteins, for purifying interacting factors and for generating antibodies against fused proteins. Analyses involving GST-tagged proteins usually require or benefit from the use of pure GST as a control or standard.
Highlights:
  • Purified from Schistosoma japonicum
  • Useful as a molecular weight marker in gel analysis of GST-tagged proteins
  • Suitable as a standard or control in ELISA and pull-down assays involving GST-tagged proteins
  • Ideal for testing tag-specific cross-reactivity of antibodies in ELISA
Specifications
Specifications
Pierce GST
GST (>/=100U/mg protein) in clear, colorless, viscous liquid, free of foreign material
10 x 1mg
Controls and tests for GST fusion protein purification
Videos