Learn More
Thermo Scientific™ Pierce™ Glutathione Superflow Agarose
Perform high-purity, large-scale purification of GST-tagged proteins with glutathione that has been immobilized to a crosslinked 6% beaded agarose.
Brand: Thermo Scientific™ 25238
Code : PN
Additional Details : Weight : 0.01000kg
Description
Perform high-purity, large-scale purification of GST-tagged proteins with Thermo Scientific™ Pierce Glutathione Superflow Agarose, which consists of glutathione that has been immobilized to a crosslinked 6% beaded agarose.
Additional Information and Online Ordering »
Highlights:
High dynamic binding capacity – binds 10mg of pure GST per mL of resin at a linear flow rate of 150cm/hour; can bind greater than 20mg/mL at 30cm/hour flow rate
High purity – provides greater than 90% purity when used to purify overexpressed GST or GST-fusion proteins from lysates
Robust – highly crosslinked 6% agarose Superflow beaded support tolerates linear flow rates up to 1200cm/hour
Compatible – maintains function after exposure to a wide variety of chemicals and pH values
Cost effective – resin is stable through multiple cycles of cleaning and reuse
Recommended for:
Medium- to large-scale FPLC purification of GST-tagged proteins
Specifications
10 mg Pure GST Binding per mL Resin at 150 cm/hour Flow Rate, 20 mg per mL at 30 cm/hour Flow Rate | |
Pierce™ | |
GST-Tagged Protein | |
Glutathione | |
Affinity, Agarose Resin |
Upon receipt store at 4°C. | |
250 mL | |
Agarose | |
Bottle |
For Research Use Only. Not for use in diagnostic procedures.