Learn More
Thermo Scientific™ Pierce™ Glutathione Magnetic Beads
Affinity purify GST-tagged fusion proteins from cell lysates using glutathione (GSH) magnetic particles for manual or robotic magnetic separation devices.
£113.04 - £380.00
Specifications
Description | Pierce Glutathione Magnetic Beads |
---|---|
Formulation | Glutathione on paramagnetic iron oxide particles (1 to 10um dia.) slurried in 20% ethyl alcohol |
Support Type | Magnetic Bead |
Sufficient For | Binding 5 to 10mg GST per mL of beads |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11824131
|
Thermo Scientific™
88821 |
4 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
11834131
|
Thermo Scientific™
88822 |
20 mL |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
Thermo Scientific™ Pierce Glutathione Magnetic Beads are superior GST-binding particles for automated magnetic separations.
Additional Information and Online Ordering »
Highlights:
High-performance beads – non-aggregating, silanized, iron oxide, paramagnetic microparticles provide exceptional uniformity for automated HTS applications
Stable affinity ligand – glutathione is covalently immobilized to particles, ensuring leach-resistance and clean purification products
High capacity – binding capacity is sufficient for both routine and demanding magnetic separation procedures
GST-tag affinity – standard glutathione-GST affinity purification chemistry in a magnetic format; bind in physiologic buffers and elute with 50mM reduced glutathione (prepared with Part No. 78259)
Properties of Glutathione Magnetic Beads:
- Composition: iron oxide particles with reduced glutathione (GSH) covalently attached to the surface and encapsulated by crosslinked agarose
- Magnetization: superparamagnetic (no magnetic memory)
- Bead size: 1 to 10μm (mean diameter)
- Concentration: 25% slurry (based on volume) in 20% ethanol
- Binding capacity: 5 to 10mg of GST fusion protein per milliliter of settled beads (i.e., 4mL of 25% slurry)
Requires:
Thermo Scientific KingFisher 96 or KingFisher Flex Instrument or other magnetic separator device
Specifications
Pierce Glutathione Magnetic Beads | |
Glutathione on paramagnetic iron oxide particles (1 to 10um dia.) slurried in 20% ethyl alcohol | |
Magnetic Bead | |
Binding 5 to 10mg GST per mL of beads |