Pierce™ Fast Semi-Dry Blotter Accessories and Replacement Parts

Perform gel-to-membrane transfer with PVDF or nitrocellulose membrane after SDS-PAGE with this semi-dry electrophoresis apparatus.

Overview
Brand: Thermo Scientific Pierce

Manufacturer Part Number: 88111

UNSPSC: 12352200

Code: PN

Additional Details:
Additional Details: Weight: 0.01000kg



Product Code. 11522531

Quantity Price
1 £ 22.85 / Pack of 4
Estimated Shipment date
from Supplier 10-04-2017
Add to basket


Description and Specification

Specification

Sufficient For Pierce Fast Semi-Dry Blotter
Item Description Banana Plug Adaptors; 2 pair
Description Banana Plugs for Pierce Fast Semi-Dry Blotter
Quantity 2 pair
Formulation Red and black adaptors for power supplies requiring thicker, metal-tipped banana terminals