Pierce™ Fast Semi-Dry Blotter Accessories and Replacement Parts

Perform gel-to-membrane transfer with PVDF or nitrocellulose membrane after SDS-PAGE with this semi-dry electrophoresis apparatus.

Overview
Brand: Thermo Scientific Pierce

Manufacturer Part Number: 88110

UNSPSC: 12352200

Code: PN

Additional Details:
Additional Details: Weight: 0.99000kg



Product Code. 10262740

Quantity Price
1 £ 129.0 / Pack of 2
Estimated Shipment date
from Supplier 11-05-2017
Add to basket


Description and Specification

Specification

Quantity 1 pair
Item Description Electrical Cables; 1 pair
Description Electical Cable for Pierce Fast Semi-Dry Blotter
Formulation Red and black (wrapped) electrical cables with plastic-tipped, banana terminals
Sufficient For Pierce Fast Semi-Dry Blotter