Learn More
Thermo Scientific™ Pierce™ Ethidium Bromide
Visualize DNA fluorescently in a variety of molecular biology applications using these dilute (0.625mg/mL) or concentrated (10mg/mL) ethidium bromide solutions.
£59.00 - £191.00
Specifications
For Use With (Application) DNA fluorescent visualization
View More Specs
Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11846714 View Documents Thermo Scientific™
17896
5mL £59.00
5mL View alternative products
Save Up to 
»
EU Stock 34 Please to purchase this item. Need a web account? Register with us today!
11856714 View Documents Thermo Scientific™
17898
5 x 10mL £191.00
Pack of 5 View alternative products
Save Up to 
»
EU Stock 10 Please to purchase this item. Need a web account? Register with us today!
Description
Description
Thermo Scientific™ Pierce Ethidium Bromide is available as both dilute (0.625mg/mL) and more concentrated (10mg/mL) solutions to accommodate most molecular biology applications for fluorescent DNA visualization.
Highlights:
0.625mg/mL EtBr Solution:
  • Each drop contains 25μg of ethidium bromide and is ideal for a 50mL agarose gel
  • After adding one drop of solution per 50mL of gel, the final concentration of EtBr will be 0.5μg/mL, which is the recommended concentration for electrophoresis of nucleic acids
  • Can also be used as the running buffer or for staining gels after electrophoresis
10mg/mL EtBr Solution:
  • A safe and convenient way to use ethidium bromide while minimizing exposure
Specifications
Specifications
DNA fluorescent visualization