Learn More
Thermo Scientific™ Pierce™ D-Luciferin, Monopotassium Salt
Get convenience and high performance at a low cost in firefly luciferase reporter assays with our D-Luciferin, formulated at greater than 99% purity as both monosodium and monopotassium salts.
Brand: Thermo Scientific™ 88294
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Thermo Scientific™ Pierce D-Luciferin is synthesized and manufactured to ensure high performance at a low cost, and the reporter is also tested by HPLC to ensure proper chirality and activity.
We are the only supplier to perform chiral-high performance liquid chromatography (HPLC) testing to ensure that the product is not only pure but also in the active form. Our D-Luciferin outshines the competition on many levels.Specifications
Store at or below -20°C. | |
ATP Assay, Gene Reporter Assay | |
Pierce™ | |
1 g | |
Chemical Substrate |
Bioluminescence | |
D-Luciferin, Monopotassium Salt | |
>99% | |
Monopotassium Salt | |
Luciferase Substrate |
For Research Use Only. Not for use in diagnostic procedures.