Thermo Scientific Pierce™ Carbonyl-reactive, 6-plex aminoxyTMT Reagents

Perform multiplexed characterization and quantitation of carbonyl-containing biomolecules by mass spectrometry (MS) with these carbonyl-reactive reagents.

Overview
Brand: Thermo Scientific

Manufacturer Part Number: 90400

UNSPSC: 41115404

Code: NEW

Additional Details:
Additional Details: Weight: 0.25000kg



Product Code. 15265753

Quantity Price
1 £ 127.0 / Pack of 6
EU Stock 56
Add to basket


Description and Specification

Specification

Kit Contents aminoxyTMT0 Label Reagent, 6 x 0.2mg
Quantity 6-rxn set
Formulation aminoxyTMT0-126 label reagent
Sufficient For Controls in 6 experiments