Learn More
Thermo Scientific™ Pierce™ Biotin-Binding Plate Sample Pack
Determine which variety of streptavidin-coated plate is best for a particular assay involving biotinylated molecules with this sample pack.
Brand: Thermo Scientific™ 15115
Code : PN
Additional Details : Weight : 0.80000kg
Description
Thermo Scientific™ Pierce Biotin-Binding Plate Sample Pack is ideal for establishing which variety of streptavidin coated plate is best for particular assay involving biotinylated molecules.
This sample pack contains streptavidin and NeutrAvidin™ Protein coated on microplates that are preblocked with either SuperBlock™, Blocking Buffer or Blocker™ BSA. This variety allows researchers to determine which plate produces the best signal-to-noise ratio for a specific experimental system. Once optimal conditions are empirically determined, the plate that provides the best results can be purchased in bulk for the remainder of the study.
Highlights:
- Two protein types: streptavidin and Thermo Scientific NeutrAvidin™ Protein
- Two blocking agents: bovine serum albumin (BSA) and Thermo Scientific SuperBlock™ Buffer
- Two coating volumes: 100μL per well and 200μL per well
Specifications
High Capacity, Standard Capacity, Affinity, Multiple | |
Fluorescence | |
Pierce | |
Coated Strip Plate | |
Streptavidin, NeutrAvidin Protein |
Store plates at 4°C. Store opened, unused plates in a resealable bag with desiccant at 4°C. Shipped at ambient temperature. | |
Sample Pack | |
Clear | |
96 | |
SuperBlock™ Buffer/Blocker™ BSA Buffer |