Learn More
Thermo Scientific™ Pierce™ AMCA Fluorescent Dyes
Label antibodies, proteins and other molecules for fluorescence imaging with these activated forms of aminomethyl coumarin acetate (AMCA) blue-fluorescent dye.
Brand:  Thermo Scientific™ 33015
Code : PN
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11879240
  • £131.17 / 5mg
Estimated Shipment
15-06-2018
Add to basket
Description
Description
AMCA (aminomethylcoumarin acetate) is a blue fluorescent dye with reactive derivatives that are used as contrasting probes for double- and triple-labeling in immunofluorescence microscopy, arrays and in situ hybridization. The desirable properties of AMCA dyes include a relatively large Stoke's shift and resistance to photobleaching. NHS-AMCA and Sulfo-NHS-AMCA are reactive towards primary amine groups on proteins, peptides and other biomolecules. AMCA-Hydrazide is used to label glycosylation sites or for conjugation to carboxyl groups using the crosslinker EDC. AMCA has an excitation maximum at 345nm and an emission maximum at 450nm.
Highlights:
 AMCA — activated derivatives of blue-fluorescent aminomethylcoumarin acetate dye
Amine-specific labeling — NHS-ester and Sulfo-NHS-ester varieties of AMCA efficiently label antibodies and other purified proteins at primary amines (lysine side chains)
Glycoprotein labeling — hydrazide variety of AMCA labels oxidized sugar groups in glycoproteins or other carbohydrates
Specifications
Specifications
Pierce Hydrazide-AMCA
Hydrazide-activated aminomethylcoumarin acetate
1 or 2 uses to label a total of 101mg of IgG using typical conditions