Learn More
Thermo Scientific™ Pierce™ ChIP-Grade Protein A/G Plus Agarose
Chromatin immunoprecipitation (chromatin IP, ChIP) assay kit containing all reagents, agarose affinity beads, and a simple, fast, reliable protocol.
£160.00 - £1510.00
Specifications
Content And Storage | Upon receipt store at 4°C |
---|---|
Form | Liquid Suspension |
Product Line | Pierce™ |
Target | IgG Antibody |
Type | Agarose |
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Quantity | Price | Quantity & Availability | ||||
10586985
|
Thermo Scientific™
26159 |
Pierce ChIP-Grade Protein A/G Plus Agarose | 0.65 mL |
£160.00
Each |
Estimated Shipment: 14-05-2024
Log in to see stock. |
||||
13474209
|
Thermo Scientific™
26161 |
ChIP-Grade Protein A/G Plus Agarose | 10 mL |
£1510.00
10mL |
Estimated Shipment: 08-05-2024
Log in to see stock. |
||||
Description
Thermo Scientific™ Pierce Agarose ChIP Kit provides a complete set of reagents and a simple, fast and reproducible protocol to perform chromatin immunoprecipitation (ChIP) assays.
This chromatin IP kit contains sufficient reagents to perform 30 ChIP assays with appropriate controls using an optimized protocol and a convenient spin-column format. The kit includes a complete Chromatin Prep Module (also available separately) to efficiently extract and solubilize crosslinked DNA-protein interaction complexes from cells, plus an immunoprecipitation module that contains high-capacity, ChIP-grade Protein A/G agarose.Highlights:
- Simple and streamlined ChIP assay protocol (< 8 hours)
- Highly efficient isolation and lysis of nuclei
- Easy and reproducible enzymatic digestion
- Low-background and high-capacity Protein A/G agarose resin
- Highly specific positive control reagents included: RNA polymerase II antibody and GAPDH PCR primers
- Fast and reproducible spin-column format; columns and collection tubes included
- High-recovery DNA purification
- Choose a complete ChIP assay kit or the separate chromatin preparation module
Includes:
Kit contains the Chromatin Prep Module (Part No. 26158; 11 components) for chromatin extraction and a complete set of immunoprecipitation reagents (15 components) for IP experiments with controls
Specifications
Upon receipt store at 4°C | |
Pierce™ | |
Agarose | |
Bottle |
Liquid Suspension | |
IgG Antibody | |
Protein A/G | |
Affinity, Agarose Resin |
For Research Use Only. Not for use in diagnostic procedures.