Learn More
Pierce™ 20X Tris-HEPES-SDS Buffer, Thermo Scientific™
Prepare Tris-HEPES-SDS running buffer required for gel electrophoresis (SDS-PAGE) with Precise Precast Gels with this space-saving stock solution.
Brand: Thermo Scientific 28368
Code : PN
Additional Details : Weight : 0.50000kg
Description
Thermo Scientific™ Pierce 20X Tris-HEPES-SDS Buffer is a space-saving stock solution used for preparing the Tris-HEPES-SDS running buffer required for gel electrophoresis (SDS-PAGE) with Thermo Scientific™ Precise and Pierce Precast Gels.
Thermo Scientific Pierce 10X and 20X Concentrated Buffers are ready to use without having to reconstitute with ultrapure water.Highlights:
Easy to use – no packets to open and no powder to dissolve
Increased accuracy – eliminates the possibility of powder remaining in a packet
Saves time – 10X or 20X concentration eliminates time spent waiting for powder to dissolve
Saves space – storage as concentrated stock minimizes bench space needed for large volume solutions
Ordering Alerts:
When diluted 20-fold in water, makes 0.1M Tris, 0.1M HEPES, 3mM SDS, pH 8
Recommended for:
Running buffer for SDS-PAGE with Thermo Scientific Precise™ and Pierce™ Precast Gels
Specifications
Pierce 20X Tris-HEPES-SDS Buffer | |
Electrophoresis | |
500 mL | |
1X = 0.1M Tris, 0.1M HEPES, 3mM SDS, pH 8 |