missing translation for 'onlineSavingsMsg'
Learn More

Picrotoxin, 98%, Thermo Scientific Chemicals

Catalog Number 10676522
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
124-87-8
C30H34O13
602.58
VJKUPQSHOVKBCO-DMRKGALJSA-N
picrotoxin
131846146
CC(=C)C1C2C3C4(C(C1C(=O)O2)(CC5C4(O5)C(=O)O3)O)C.CC12C3C4C(C(C1(CC5C2(O5)C(=O)O3)O)C(=O)O4)C(C)(C)O
124-87-8
C30H34O13
602.58
VJKUPQSHOVKBCO-DMRKGALJSA-N
picrotoxin
131846146
CC(=C)C1C2C3C4(C(C1C(=O)O2)(CC5C4(O5)C(=O)O3)O)C.CC12C3C4C(C(C1(CC5C2(O5)C(=O)O3)O)C(=O)O4)C(C)(C)O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 199.0°C to 202.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (Mixture of Picrotoxinin and Picrotin) (HPLC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15,7527
Specific Rotation − 32.00
Solubility Information Solubility in water: 1g/5mL (100°C). Other solubilities: soluble in boiling alcohol (1gr/3mL),dilute acids,and alkalies; sparingly soluble in ether and ccl4,readily soluble in strong ammonia water and in aq.,solns of naoh.,1gr/13.5 mL in 95% ethan
Formula Weight 602.58
Percent Purity 98%
Physical Form Crystals or Powder
Chemical Name or Material Picrotoxin
Show More Show Less
compliance-icons
Product Identifier
  • Picrotoxin
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
Hazard Statement
  • H300-Fatal if swallowed.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'