missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Picrolonic acid, 98+%

Brand:  Alfa Aesar™ A10836.14

Code : NEW

Additional Details : CAS Number : 550-74-3 Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11409802

  • £150.10 / 25g


Estimated Shipment date
from Supplier 31-05-2018
Add to basket

Description

Description

Applications
Reagent for the isolation of amines as crystalline π-complexes.Picrolonic acid is an analytical reagent used for the estimation of calcium, magnesium, lead, strontium and thorium. It is also used for the analysis of alkaloids, tryptophan and phenylalanine.

Solubility
Slightly soluble in water. Soluble in alcohol.

Notes
Incompatible with strong oxidizing agents and strong bases.
Specifications

Chemical Identifiers

Picrolonic acid
C10H8N4O5
MFCD00003133
14,7412
550-74-3
264.21
623723
3-Methyl-4-nitro-1-(4-nitrophenyl)-2-pyrazolin-5-one; Picrolic acid

Specifications

Picrolonic acid
25g
≥98%
264.21
623723
3-Methyl-4-nitro-1-(4-nitrophenyl)-2-pyrazolin-5-one; Picrolic acid
≈120°C (dec.)
550-74-3
C10H8N4O5
MFCD00003133
14,7412
Videos
SDS