Learn More
Honeywell Safety Products™ Resistop Gloves
Comfortable and sturdy polyamide-cotton knit construction gives these gloves thermal and mechanical resistance. Honeywell Safety Products™ Resistop Gloves are non-pilling and suitable for precision work.
Brand: Honeywell Safety Products™ RGT099T9
Code : NEW
Additional Details : Weight : 0.08550kg
Description
- Suitable for handling sharp and hot objects.
- Non-pilling knit polyamide exterior.
- Abrasion, cutting, thermal resistance.
- Suitable for precision work.
- Conforms to hand.
- Cotton interior for comfort and sweat absorption.
- Thick construction absorbs shocks.
- White color enhances hygiene and cleanliness.
Specifications
Dexterity: 5 | |
Handling of sharp parts in automotive and mechanical industries. Assembling operations, removal of molds up to 100°C for 30 seconds maximum. Automotive, Aviation, Glass, Logistics and Offshore industries. | |
Polyamide, Cotton | |
9 | |
10Pair/PK | |
CATEGORY CE II Intermediary risks, EN 420-03 EEC Basic standards, EN 388-03 Mechanical hazards, EN 407-2004 Thermal hazards, EC Category PPE 2, EC Certificate Number N°0075/797/162/09/08/0345, EC Attestation Number N°0075/797/162/09/08/0345 |
Knitted Polyamide/Cotton | |
6cm | |
Cuts,Heat | |
4 | |
White |