missing translation for 'onlineSavingsMsg'
Learn More

Piceatannol, 96%, ACROS Organics™

Brand:  Acros Organics 328450100

UNSPSC : 12350000

Code : 29

Additional Details : CAS Number : 10083-24-6 Weight : 0.05001kg

Quantity 10mg

 View more versions of this product

Product Code. 10173481

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Piceatannol
≥95%
C14H12O4
MFCD00221715
10083-24-6
96%
244.3
3,4,3',5'-Tetrahydroxy-trans-stilbene

Specifications

Piceatannol
10mg
Authentic
≥95%
C14H12O4
MFCD00221715
Yellow to golden or green
10083-24-6
Crystalline Powder
96%
244.3
3,4,3',5'-Tetrahydroxy-trans-stilbene
Documentation

Technical Documents