missing translation for 'onlineSavingsMsg'
Learn More

PI 828, Tocris Bioscience™

PI 3-kinase inhibitor, more potent than LY 294002 (Cat. No. 1130)

Brand:  Tocris (Bio-Techne) 281410

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13441884

  • £188.76 / 10mg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

PI 3-Kinase inhibitor (IC50 values are 0.098, 0.183, 0.227 and 1.967 μM for p110β, p110α, p110δ and p110γ respectively) that displays higher potency than LY 294002 (Cat No. 1130). Can be immobilized onto a solid phase.
Specifications

Chemical Identifiers

942289-87-4
322.364
8-4-aminophenyl-2-morpholino-4h-chromen-4-one, ly 294002, 4gamma-nh2, 2-4-morpholinyl-8-4-aminopheny l-4h-1-benzopyran-4-one, 8-4-aminophenyl-2-4-morpholinyl-4h-1-benzopyran-4-one, 8-4-aminophenyl-2-morpholin-4-yl-4h-1-benzopyran-4-one, 8-4-aminophenyl-2-morpholin-4-yl chromen-4-one
8-(4-aminophenyl)-2-morpholin-4-ylchromen-4-one
C19H18N2O3
WUKMIBOGGXMBAC-UHFFFAOYSA-N
25181195
C1COCCN1C2=CC(=O)C3=C(O2)C(=CC=C3)C4=CC=C(C=C4)N

Specifications

PI 828
942289-87-4
8-4-aminophenyl-2-morpholino-4h-chromen-4-one, ly 294002, 4gamma-nh2, 2-4-morpholinyl-8-4-aminopheny l-4h-1-benzopyran-4-one, 8-4-aminophenyl-2-4-morpholinyl-4h-1-benzopyran-4-one, 8-4-aminophenyl-2-morpholin-4-yl-4h-1-benzopyran-4-one, 8-4-aminophenyl-2-morpholin-4-yl chromen-4-one
C1COCCN1C2=CC(=O)C3=C(O2)C(=CC=C3)C4=CC=C(C=C4)N
322.364
322.36
10mg
C19H18N2O3
WUKMIBOGGXMBAC-UHFFFAOYSA-N
8-(4-aminophenyl)-2-morpholin-4-ylchromen-4-one
25181195
>99%
Videos