missing translation for 'onlineSavingsMsg'
Learn More

uvex Phynomic Pro Safety Gloves

Product Code. 17220381 Shop All Uvex Products
Click to view available options
Size:
6
7
8
9
10
11
12
Unit Size:
10 pairs
This item is not returnable. View return policy

Product Code. 17220381

Brand: uvex 6006209

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Second skin design is light and flexible to help minimize excessive perspiration, while aqua-polymer foam coating repels dirt and moisture.

  • Lightweight construction acts as a second-skin for tactile sensitivity
  • Bamboo/polyamide/elastane liner provides a high level of user comfort
  • Natural fibers help to absorb water vapor
  • Aqua-polymer foam coating repels dirt and moisture
  • Breathable materials help prevent excessive perspiration
  • Enhanced grip in dry, damp, and oily applications
  • 3D Ergo technology ensures a secure fit
  • Free of harmful substances in accordance with OEKO-TEX™ Standard 100
  • Skin tolerance confirmed by the proDERM Institute for Applied Dermatological Research (11.0356-02, 11.0482-11)
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance OEKO-TEX™ Standard 100
Applications Industrial
Coating Material Aqua-Polymer Pro
Color Blue, Gray
Exterior Finish Smooth
Hand Ambidextrous
Latex Free Yes
Material Bamboo, Polyamide, Elastane
Product Type Mechanical Protection Gloves
Size 9
EN388 Standard 2121X
For Use With (Application) Assembly, Construction, Maintenance, Outdoor Activities, and Repairs
Gender Unisex
Glove Cuff Style Knit
Series Phynomic Pro
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.