Learn More
Phycoerythrin Monoclonal Antibody (eBioPE-DLF), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-4120-85
137.34 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Description: The eBioPE-DLF monoclonal antibody reacts with phycoerythrin (PE), PE-Cy5 and PE-Cy7, with no crossreactivity to fluorescein (FITC) or allophycocyanin (APC). When using eBioPE-DLF as a second-step antibody, in combination with an appropriate third step, to bind a PE-conjugated primary antibody, there is no decrease in the fluorescent intensity of the directly conjugated PE antibody. Therefore no interference or quenching of the PE fluorochrome is observed. The eBioPE-DLF can be used for separation of cells labeled with PE-conjugated antibodies or staining. Please note that the optimal concentration for this antibody will vary depending on the density of the primary PE-conjugated antibody and the application tested. Applications Reported: This eBioPE-DLF antibody has been reported for use in flow cytometric analysis. Applications Tested: This eBioPE-DLF antibody has been tested by flow cytometric analysis. This can be used at less than or equal to 0.125 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE. Aggregation: Less than 10%, as determined by HPLC. Filtration: 0.2 μm post-manufacturing filtered.
Anti-RPE (R-Phycoerythrin) antibodies can be used to bind conjugated primary antibody without decreasing fluorescent intensity of the RPE fluorochrome. This method is used for separation of cells labeled with RPE-conjugated antibodies.Specifications
Phycoerythrin | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
Mouse | |
500 μg | |
Primary | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
eBioPE-DLF | |
Unconjugated | |
PE; RPE; R-PE; R-phycoerythrin | |
Affinity chromatography | |
RUO | |
4° C | |
Liquid | |
Tag |
For Research Use Only.