Thermo Scientific™ Phusion™ U Multiplex PCR Master Mix PROMO
High performance multiplex PCR master mix for fast and reliable simultaneous amplification of multiple targets
Overview
Brand: Thermo Scientific™
Manufacturer Part Number: F-562L-RMP6225
500RXN Phusion U Multiplex PCR Master Mix, 500 x 50 µL rxns
UNSPSC: 12352204
Additional Details:
Additional Details: Weight: 0.08000kg
Product Code. 15593511
Quantity Price
1 £ 702.0 / 500 reactions
Estimated Shipment date
from Supplier 10-03-2017
Add to basket
Description and Specification
Thermo Scientific Phusion U PCR Master Mix is a ready-to-use, 2X end-point PCR master mix designed for simultaneous amplification of multiple targets in a single tube. Over 20 primer pairs may be combined into a single reaction for highly specific and efficient multiplexing over a broad range of primer and template concentrations.
The master mix is based on Phusion U Hot Start DNA Polymerase, a high performance enzyme developed using fusion technology. Similarly to other enzymes from Phusion family, Phusion U DNA Polymerase incorporates more nucleotides per binding event than any other DNA polymerase. This allows achieving high yields of PCR products with shorter extention times and consequently reduced total protocol times. Significantly enhanced processivity also enables successful multiplex PCR on difficult targets such as GC-rich templates or templates of suboptimal purity. Being highly tolerant of many PCR inhibitors, Phusion U Multiplex PCR Master Mix can even be used with unpurified samples such as blood or serum.
Specificity of multiplex PCR is increased by Affibody-mediated hot start mechanism. Reversibly bound Affibody ligand inhibits the polymerase at ambient temperatures preventing amplification of nonspecific products and formation of primer-dimers.
The Phusion U Multiplex PCR Master Mix contains a proprietary buffer with balanced concentrations of all PCR components eliminating the need for tedious optimization. The Phusion U Green Multiplex PCR Master Mix further simplifies the workflow - it includes a density reagent and two electrophoresis tracking dyes for direct loading of PCR products on gels.
Highlights
  • Specific and efficient multiplex PCR of over 20 targets up to 2.5 kb
  • Excellent results with templates of suboptimal purity due to inhibitor tolerance
  • Fast cycling and direct loading of PCR products on gels
  • Applications
    • Genotyping
    • Pathogen detection
    • Food testing
    • Analysis of genetically modified organisms
    • Amplification of microsatellites
    Includes
    Phusion U and Phusion U Green Multiplex PCR Master Mixes contain Phusion U Hot Start DNA Polymerase, 2X reaction buffer, 3 mM MgCl2, 400 °m of each dATP, dTTP, dCTP and dGTP.
    The master mixes are supplied with nuclease-free water.