Thermo Scientific™ Phusion™ Hot Start II High-Fidelity DNA Polymerase PROMO

The most accurate hot start DNA polymerase on the market featuring extreme specificity and improved performance.

Overview
Brand: Thermo Scientific™

Manufacturer Part Number: F-565S-RMP6225

UNSPSC: 41106300

Additional Details:
Additional Details: Weight: 0.02000kg



Product Code. 15543521

Quantity Price
1 £ 149.0 / 100 reactions
Estimated Shipment date
from Supplier 21-04-2017
Add to basket


Description and Specification

Phusion Hot Start II High Fidelity DNA Polymerase combines the Phusion DNA Polymerase and a reversibly bound, specific Affibody ligand. Affibody inhibits the DNA polymerase activity at room temperature and thus prevents the amplification of nonspecific products.

The Affibody ligand also inhibits the 3‡→5‡ exonuclease activity of the polymerase, preventing degradation of primers and template DNA during reaction set up. At polymerization temperatures, the ligand is released, rendering the polymerase fully active. Phusion Hot Start II DNA Polymerase does not require a separate activation step in the PCR protocol as it is immediately reactivated at high temperatures.

New Phusion Green Hot Start II High-Fidelity DNA Polymerase is a combination of Phusion Hot Start II DNA Polymerase and 5X Phusion Green Buffers. The buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with excelent performance of Phusion Hot Start II DNA Polymerase and is compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.

Thermo Scientific Phusion Hot Start II High Fidelity DNA Polymerase is the most accurate hot start DNA polymerase on the market. It combines the Phusion DNA Polymerase and a reversibly bound, specific Affibody ligand. Affibody inhibits the DNA polymerase activity at room temperature and thus prevents the amplification of nonspecific products.
The Affibody ligand also inhibits the 3'→5' exonuclease activity of the polymerase, preventing degradation of primers and template DNA during reaction set up. At polymerization temperatures, the ligand is released, rendering the polymerase fully active. Phusion Hot Start II DNA Polymerase does not require a separate activation step in the PCR protocol as it is immediately reactivated at high temperatures.
New Phusion Green Hot Start II High-Fidelity DNA Polymerase is a combination of Phusion Hot Start II DNA Polymerase and 5X Phusion Green Buffers. The buffers include a density reagent and two tracking dyes for direct loading of PCR products on a gel. The colored buffer does not interfere with excelent performance of Phusion Hot Start II DNA Polymerase and is compatible with downstream applications such as DNA sequencing, ligation and restriction digestion.


Highlights
• Reaction set up can be done at room temperature
• Reduces non-specific amplification
• Prevents primer degradation during set up
• Zero-time reactivation due to unique hot start technology
• Extreme fidelity (52X Taq)
• Speed of the polymerase allows short extension times
• Robust reactions, minimal optimization needed
• Increased product yields with minimal enzyme amounts
• Direct loading on gel with Green Buffer
Applications
• High-fidelity PCR
• High throughput
• Difficult (GC-rich) templates
• Template generation for sequencing
• Multiplex PCR
• Long-range PCR
• Cloning
• Mutagenesis†
• Microarray
†<a href="/mutagenesis/phusion-site-directed-mutagenesis-kit/">Phusion Site-Directed Mutagenesis Kit</a> also available.
Includes
Phusion Hot Start II High-Fidelity DNA Polymerase
• Phusion Hot Start II High-Fidelity DNA Polymerase (2 U/μL)
• 5X Phusion HF Buffer
• 5X Phusion GC Buffer
• DMSO
• 50 mM MgCl2 solution
Phusion Green Hot Start II High-Fidelity DNA Polymerase
• Phusion Hot Start II High-Fidelity DNA Polymerase (2 U/μL)
• 5X Phusion Green HF Buffer
• 5X Phusion Green GC Buffer
• DMSO
• 50 mM MgCl2 solution
Both Phusion and Phusion Green Buffers provide 1.5 mM MgCl2 in the final 1X concentration.

Hazardous:No
> Shelf Life:24 Months
> Shipping Condition:Dry Ice
> Storage buffer:20 mM Tris-HCl (pH 7.4 at 25°C), 0.1 mM EDTA, 1 mM DTT, 100 mM KCl, stabilizers, 200 μg/mL BSA, and 50% glycerol.
> Storage Condition:-20°C