Thermo Scientific™ Phusion Hot Start High-Fidelity DNA Polymerase

Increase the specificity of PCR amplification by preventing non-specific extension with this highly accurate DNA polymerase, now available with a hot start modification.

Overview
Brand: Thermo Scientific™

Manufacturer Part Number: F-540L

PHUSION HOT START DNA POL 500U

UNSPSC: 12352200

Code: 87

Additional Details:
Additional Details: Weight: 1.71460kg



Product Code. 10004397

Quantity Price
1 £ 487.0 / 500 units
EU Stock 20
Add to basket


Description and Specification

Specification

Storage Requirements Recommended storage temperature -20°C. Stable for one year from the assay date.
Description Phusion™ Hot Start High-Fidelity DNA Polymerase
Quantity 500U (2U/µL)
For Use With (Application) Fast PCR, high-fidelity PCR, cloning, long or difficult amplicons, DHPLC, microarray
Includes Phusion Hot Start High-Fidelity DNA Polymerase is supplied with 5x Phusion HF Buffer, 5x Phusion GC Buffer, DMSO and 50 mM MgCl2 solution. Both Phusion HF Buffer and Phusion GC Buffer provide 1.5 mM MgCl2 in the final 1x concentration.

Storage buffer

20 mM Tris-HCl (pH 7.4 at 25°C), 0.1 mM EDTA, 1 mM DTT, 100 mM KCl, stabilizers, 200 µg/ml BSA and 50% glycerol.

Storage

Recommended storage temperature -20°C.