Thermo Scientific™ Phusion High-Fidelity DNA Polymerase PROMO
Get superior performance for cloning and other applications requiring high fidelity with one of the most accurate thermostable DNA polymerases available.
Overview
Brand: Thermo Scientific™
Manufacturer Part Number: F-532S-RMP6225
UNSPSC: 41106300
Additional Details:
Additional Details: Weight: 0.00900kg
Product Code. 15543501
Quantity Price
1 £ 134.0 / 100 reactions
Estimated Shipment date
from Supplier 03-03-2017
Add to basket
Description and Specification
Highlights
  • Extreme fidelity (error rate 4.4 x 10-7 in Phusion HF Buffer)
  • Polymerase speed allows short extension times (15 to 30 s/kb)
  • Robust performance, minimal optimization needed
  • Increased product yields with minimal enzyme amounts
  • Direct loading on gel with Green Buffer