Thermo Scientific™ Phusion™ Flash High-Fidelity PCR Master Mix PROMO
Perform short PCR protocols without compromising fidelity or yield with this master mix.
Overview
Brand: Thermo Scientific™
Manufacturer Part Number: F-548L-RMP6225
500RXN Phusion Flash High-Fidelity PCR Master Mix, 500 x 20 µL rxns
UNSPSC: 41106300
Additional Details:
Additional Details: Weight: 0.00900kg
Product Code. 15513511
Quantity Price
1 £ 253.0 / 500 reactions
Estimated Shipment date
from Supplier 30-01-2017
Add to basket
Description and Specification
Highlights
  • Extreme speed: extension times of 15 s/kb or less
  • Hot start modification allowing “zero-time reactivation”
  • Accuracy: proofreading DNA polymerase with a fidelity of 25X Taq
  • High yields in reduced time