missing translation for 'onlineSavingsMsg'
Learn More

Phthaloyl chloride, 90%, AcroSeal™, Thermo Scientific™

Catalog Number 13525000
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass bottle
This item is not returnable. View return policy
88-95-9
FYXKZNLBZKRYSS-UHFFFAOYSA-N
phthaloyl chloride, phthaloyl dichloride, phthalic chloride, phthalyl chloride, phthalic dichloride, phthalic acid dichloride, phthalyl dichloride, 1,2-benzenedicarbonyl dichloride, o-phthaloyl dichloride, phthaloyldichloride
6955
benzene-1,2-dicarbonyl chloride
C1=CC=C(C(=C1)C(=O)Cl)C(=O)Cl
This item is not returnable. View return policy
88-95-9
FYXKZNLBZKRYSS-UHFFFAOYSA-N
phthaloyl chloride, phthaloyl dichloride, phthalic chloride, phthalyl chloride, phthalic dichloride, phthalic acid dichloride, phthalyl dichloride, 1,2-benzenedicarbonyl dichloride, o-phthaloyl dichloride, phthaloyldichloride
6955
benzene-1,2-dicarbonyl chloride
C1=CC=C(C(=C1)C(=O)Cl)C(=O)Cl

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point 12.0°C
Density 1.4000g/mL
Boiling Point 269.0°C to 271.0°C
Flash Point 113°C
Assay Percent Range 88% min
Packaging AcroSeal™ Glass bottle
Molecular Formula C8H4Cl2O2
Linear Formula C6H4(COCl)2
Refractive Index 1.5674 to 1.5694
Quantity 800 mL
Beilstein 09,805
Fieser 01,882; 14,263
Merck Index 15,7487
Specific Gravity 1.4
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in ether, decomposes in alcohol
Molecular Weight (g/mol) 203.02
Formula Weight 203.02
Percent Purity 90%
Chemical Name or Material Phthaloyl chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'