missing translation for 'onlineSavingsMsg'
Learn More

Phthalimide, 99%, Thermo Scientific Chemicals

Catalog Number p-3593491
12.65 GBP valid until 2024-12-31
Use promo code "22914" to get your promotional price.


Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Plastic bottle
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
85-41-6
C8H5NO2
147.13
MFCD00005881
XKJCHHZQLQNZHY-UHFFFAOYSA-N
phthalimide, 1h-isoindole-1,3 2h-dione, o-phthalimide, isoindoline-1,3-dione, o-phthalic imide, 1,3-isoindolinedione, benzoimide, 2-diazoindan-1,3-dione, phenylimide, phthalimid
6809
CHEBI:38817
isoindole-1,3-dione
O=C1NC(=O)C2=CC=CC=C12
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
85-41-6
C8H5NO2
147.13
MFCD00005881
XKJCHHZQLQNZHY-UHFFFAOYSA-N
phthalimide, 1h-isoindole-1,3 2h-dione, o-phthalimide, isoindoline-1,3-dione, o-phthalic imide, 1,3-isoindolinedione, benzoimide, 2-diazoindan-1,3-dione, phenylimide, phthalimid
6809
CHEBI:38817
isoindole-1,3-dione
O=C1NC(=O)C2=CC=CC=C12

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >233.5°C
Color White to Yellow
Boiling Point 366.0°C
Flash Point 165°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (HPLC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 21, 458
Fieser 07,330
Merck Index 15, 7485
Solubility Information Solubility in water: 0.4-0.6g/L (20°C). Other solubilities: 5 g/100g boiling alcohol, freely soluble in aqueous alkali hydroxides, soluble in boiling acetic acid, acetone, hot ether, and dimethyl sulfoxide, almost insoluble in benzene, petroleum ether
Sublimation Point 0366.0°C
Formula Weight 147.13
Percent Purity 99%
Physical Form Crystalline Flakes
Chemical Name or Material Phthalimide
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'