missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phthalic Anhydride, 99+%, Extra Pure, SLR, Fisher Chemical™
Click to view available options
Quantity:
500 g
1 kg
Specifications
Packaging | Plastic powder jar |
Quantity | 500 g |
Color | White |
Formula Weight | 148.12g/mol |
Vapor Pressure | 0.01mbar at 20°C |
pH | 2 |
Melting Point | 129°C |
Boiling Point | 284°C |
Physical Form | Solid |
Safety and Handling
Product Identifier
- Phthalic anhydride
- Danger
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'