missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Phosphorus(V) oxychloride, 99.999% (metals basis)

Brand:  Alfa Aesar™ 011847.06

Code : NEW

Additional Details : CAS Number : 10025-87-3 Weight : 0.00500kg Transport : UN number : 1810 Chem class : 1a Pack group : I

Quantity 5g

 View more versions of this product

Product Code. 15461007

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »


Specifications

Chemical Identifiers

Phosphorus(V) oxychloride
POCl3
MFCD00011443
14,7349
10025-87-3
153.33
UN1810
Phosphorus(V) trichloride oxide; Phosphoryl chloride

Specifications

Phosphorus(V) oxychloride
Pungent
105.8°C
1.25°C
10025-87-3
Liquid
(metals basis)
153.33
UN1810
Phosphorus(V) trichloride oxide; Phosphoryl chloride
1.645g/mL
Moisture Sensitive
Colorless
5g
1.460
99.999%
POCl3
MFCD00011443
14,7349
Videos
SDS