missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus(V) oxychloride, 99.995% (metals basis), Thermo Scientific Chemicals

Catalog Number 15405995
Click to view available options
Quantity:
25 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
10025-87-3
Cl3OP
153.323
MFCD00011443
XHXFXVLFKHQFAL-UHFFFAOYSA-N
phosphorus oxychloride, phosphoryl chloride, phosphoric trichloride, phosphoryl trichloride, phosphoroxychloride, trichlorophosphine oxide, phosphorus oxytrichloride, fosforoxychlorid, phosphorous oxychloride, phosphorylchlorid
24813
CHEBI:30336
O=P(Cl)(Cl)Cl
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
10025-87-3
Cl3OP
153.323
MFCD00011443
XHXFXVLFKHQFAL-UHFFFAOYSA-N
phosphorus oxychloride, phosphoryl chloride, phosphoric trichloride, phosphoryl trichloride, phosphoroxychloride, trichlorophosphine oxide, phosphorus oxytrichloride, fosforoxychlorid, phosphorous oxychloride, phosphorylchlorid
24813
CHEBI:30336
O=P(Cl)(Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 105.8°C
Melting Point 1.25°C
Color Colorless
Physical Form Liquid
Quantity 25 g
Linear Formula POCl3
UN Number UN1810
Merck Index 14,7349
Solubility Information Insoluble in organic solvents and reacts with water.
Formula Weight 153.33
Percent Purity 99.999%
Odor Pungent
Refractive Index 1.460
Sensitivity Moisture sensitive
Density 1.645g/mL
Assay (metals basis)
Chemical Name or Material Phosphorus(V) oxychloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'