missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus(V) chloride, 98%, Thermo Scientific Chemicals

Catalog Number 11356038
Click to view available options
Quantity:
100 g
500 g
4 x 500 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
10026-13-8
Cl5P
208.224
MFCD00011439
UHZYTMXLRWXGPK-UHFFFAOYSA-N
phosphorus pentachloride, phosphorus v chloride, pentachlorophosphorane, phosphorane, pentachloro, pcl5, phosphorous pentachloride, phosphorpentachlorid, phosphoric perchloride, phosphorus perchloride, unii-0ex753tydu
24819
CHEBI:30335
pentachloro-$l^{5}-phosphane
P(Cl)(Cl)(Cl)(Cl)Cl
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
10026-13-8
Cl5P
208.224
MFCD00011439
UHZYTMXLRWXGPK-UHFFFAOYSA-N
phosphorus pentachloride, phosphorus v chloride, pentachlorophosphorane, phosphorane, pentachloro, pcl5, phosphorous pentachloride, phosphorpentachlorid, phosphoric perchloride, phosphorus perchloride, unii-0ex753tydu
24819
CHEBI:30335
pentachloro-$l^{5}-phosphane
P(Cl)(Cl)(Cl)(Cl)Cl

For Use As Dehydrating and Chlorinating Agent, Catalyst

Phosphorus(V) chloride can be used for both oxidative and substitutive chlorinations, & for both organic and inorganic compounds. Dehydrating agent. Precursor to Vilsmeier reagent, lithium hexafluorophosphate electrolyite for lithium batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 162° (sublimation)
Color Yellow
Physical Form Solid
Quantity 4 x 500 g
Assay Percent Range 98%
Solubility Information Soluble in carbon disulfide and carbon tetrachloride.
Formula Weight 208.24
Odor Pungent
Sensitivity Moisture sensitive
Appearance Yellowish crystals
Vapor Pressure 0.02 hPa at 20°C
Density 2.114
Chemical Name or Material Phosphorus(V) chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'