missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphorus pentachloride, 98%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
1 kg
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent
Specifications
Color | Yellow |
Physical Form | Crystalline Powder and Scales |
Quantity | 100 g |
Assay Percent Range | 98% |
Linear Formula | PCl5 |
Fieser | 01,866; 04,388; 05,534; 07,290 |
SMILES | P(Cl)(Cl)(Cl)(Cl)Cl |
IUPAC Name | pentachloro-$l^{5}-phosphane |
Molecular Weight (g/mol) | 208.224 |
Formula Weight | 208.24 |
Show More |
Safety and Handling
Product Identifier
- Phosphorus pentachloride
- Danger
- Acute toxicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
- H302-Harmful if swallowed.
- H330-Fatal if inhaled.
- H314-Causes severe skin burns and eye damage.
- H373-May cause damage to organs.
- EUH014-Reacts violently with water.
- EUH029-Contact with water liberates toxic gas.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P402+P404-Store in a dry place. Store in a closed container.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'