missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus oxychloride, 99%, Thermo Scientific Chemicals

Catalog Number 10032470
Click to view available options
Quantity:
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
10025-87-3
MFCD00011443
XHXFXVLFKHQFAL-UHFFFAOYSA-N
phosphorus oxychloride, phosphoryl chloride, phosphoric trichloride, phosphoryl trichloride, phosphoroxychloride, trichlorophosphine oxide, phosphorus oxytrichloride, fosforoxychlorid, phosphorous oxychloride, phosphorylchlorid
24813
CHEBI:30336
O=P(Cl)(Cl)Cl
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Sea freight only: For EU stock allow up to 15 days delivery.
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
10025-87-3
MFCD00011443
XHXFXVLFKHQFAL-UHFFFAOYSA-N
phosphorus oxychloride, phosphoryl chloride, phosphoric trichloride, phosphoryl trichloride, phosphoroxychloride, trichlorophosphine oxide, phosphorus oxytrichloride, fosforoxychlorid, phosphorous oxychloride, phosphorylchlorid
24813
CHEBI:30336
O=P(Cl)(Cl)Cl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

C-X bond formation reagent, Oxidation reagent
TRUSTED_SUSTAINABILITY
Physical Form Liquid
Quantity 250 g
Assay Percent Range 99%
Molecular Formula Cl3OP
Linear Formula POCl3
Fieser 01,876; 02,330; 03,228; 04,390; 05,535; 07,292; 09,374; 11,429; 13,249; 15,267; 17,288
Merck Index 15, 7461
Molecular Weight (g/mol) 153.323
Formula Weight 153.33
Percent Purity ≥98.5% (ex Cl)
Packaging Glass bottle
Refractive Index 1.4600 to 1.4620 (20°C, 589nm)
Density 1.6450g/mL
Chemical Name or Material Phosphorus oxychloride
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Phosphorus oxychloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H330-Fatal if inhaled.
  • H314-Causes severe skin burns and eye damage.
  • H372-Causes damage to organs.
  • H290-May be corrosive to metals.
  • EUH029-Contact with water liberates toxic gas.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'