missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphorus oxychloride, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagent, Oxidation reagent
Specifications
Physical Form | Liquid |
Quantity | 250 g |
Assay Percent Range | 99% |
Molecular Formula | Cl3OP |
Linear Formula | POCl3 |
Fieser | 01,876; 02,330; 03,228; 04,390; 05,535; 07,292; 09,374; 11,429; 13,249; 15,267; 17,288 |
Merck Index | 15, 7461 |
Molecular Weight (g/mol) | 153.323 |
Formula Weight | 153.33 |
Percent Purity | ≥98.5% (ex Cl) |
Show More |
Safety and Handling
Product Identifier
- Phosphorus oxychloride
- Danger
- Acute toxicity Category 2
- Skin corrosion/irritation Category 1 A
- Serious eye damage/eye irritation Category 1
- Specific target organ toxicity Category 1
- Substances and mixtures corrosive to metals Category 1
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- H330-Fatal if inhaled.
- H314-Causes severe skin burns and eye damage.
- H372-Causes damage to organs.
- H290-May be corrosive to metals.
- EUH029-Contact with water liberates toxic gas.
- EUH014-Reacts violently with water.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P310-Immediately call a POISON CENTER/doctor/.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'