missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Phosphorus(III) chloride, 99.997% (metals basis)

Brand:  Alfa Aesar™ 022988.22

Code : NEW

Additional Details : CAS Number : 7719-12-2 Weight : 0.10000kg Transport : UN number : 1809 Chem class : 6.1 Pack group : I

Quantity 100g

 View more versions of this product

Product Code. 15456065

  • £431.70 / 100g


Estimated Shipment date
from Supplier 26-06-2018
Add to basket

Specifications

Chemical Identifiers

Phosphorus(III) chloride
137.33
UN1809
7719-12-2
MFCD00011438
14,7358

Specifications

Phosphorus(III) chloride
Acrid
76°C
100g
1.516
99.997%
137.33
UN1809
1.574g/mL at 21°C
Moisture Sensitive
−111.8°C
7719-12-2
Liquid
(metals basis)
MFCD00011438
14,7358
Videos
SDS