missing translation for 'onlineSavingsMsg'
Learn More

Phosphorus(III) chloride, 98% min, Thermo Scientific Chemicals

Catalog Number 15411997
Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7719-12-2
Cl3P
137.32
MFCD00011438
FAIAAWCVCHQXDN-UHFFFAOYSA-N
phosphorus trichloride, phosphorous trichloride, trichlorophosphine, phosphorus chloride, phosphorous chloride, phosphorus iii chloride, pcl3, phosphorus chloride pcl3, phosphine, trichloro, fosfortrichloride
24387
CHEBI:30334
trichlorophosphane
P(Cl)(Cl)Cl
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
7719-12-2
Cl3P
137.32
MFCD00011438
FAIAAWCVCHQXDN-UHFFFAOYSA-N
phosphorus trichloride, phosphorous trichloride, trichlorophosphine, phosphorus chloride, phosphorous chloride, phosphorus iii chloride, pcl3, phosphorus chloride pcl3, phosphine, trichloro, fosfortrichloride
24387
CHEBI:30334
trichlorophosphane
P(Cl)(Cl)Cl

Phosphorus(III) chloride is used as a reagent in organic synthesis. It is utilized in the conversion of primary alcohol and secondary alcohol to alkyl chlorides. It acts as a precursor to phosphorous pentachloride, phosphorous oxychloride, thiophosphoryl chloride and triphenylphosphine. It is also used to prepare phosphite esters, which is used Horner-Wadsworth-Emmons reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Boiling Point 76°C
Melting Point -111.8°C
Color Yellow
Physical Form Liquid
Quantity 250 g
Assay Percent Range 98% min.
UN Number 1809
Solubility Information Miscible with benzene,carbon disulfide,ether,chloroform and carbon tetrachloride.
Formula Weight 137.33
Odor Acrid
Refractive Index 1.516
Vapor Pressure 95mm Hg at 20°C
Density 1.574 g/cm3 at 20°C
Chemical Name or Material Phosphorus(III) Chloride
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'