Learn More
Phosphorous acid, 98+%, Thermo Scientific Chemicals
Use promo code "21615" to get your promotional price.
Description
Phosphorus acid is used as an intermediate in the preparation of other phosphorus compounds like potassium phosphite, ammonium phosphite and calcium phosphite. It is actively involved in the preparation of phosphites like aminotris(methylenephosphonic acid) (ATMP), 1-hydroxyethane 1,1-diphosphonic acid (HEDP) and 2-phosphonobutane-1,2,4-tricarboxylic Acid (PBTC), which find application in water treatment as a scale or corrosive inhibitor. It is also used in chemical reactions as a reducing agent. Its salt, lead phosphite is used as PVC stabilizer. It is also used as precursor in the preparation of phosphine as well as an intermediate in the preparation of other phosphorus compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phosphorus acid is used as an intermediate in the preparation of other phosphorus compounds like potassium phosphite, ammonium phosphite and calcium phosphite. It is actively involved in the preparation of phosphites like aminotris(methylenephosphonic acid) (ATMP), 1-hydroxyethane 1,1-diphosphonic acid (HEDP) and 2-phosphonobutane-1,2,4-tricarboxylic Acid (PBTC), which find application in water treatment as a scale or corrosive inhibitor. It is also used in chemical reactions as a reducing agent. Its salt, lead phosphite is used as PVC stabilizer. It is also used as precursor in the preparation of phosphine as well as an intermediate in the preparation of other phosphorus compounds.
Solubility
Soluble in water and alcohol.
Notes
Air sensitive. Hygroscopic. Incompatible with strong bases, oxidizing agents and metals.
Specifications
Melting Point | 74°C |
Boiling Point | 200°C (decomposition) |
Density | 1.651 |
Odor | Odorless |
Sensitivity | Hygroscopic |
Quantity | 250 g |
UN Number | UN2834 |
Merck Index | 14,7346 |
Solubility Information | Soluble in water and alcohol. |
Formula Weight | 82 |
Show More |
RUO – Research Use Only