missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric acid, TraceSELECT™ Ultra, Honeywell Fluka™
Click to view available options
Quantity:
250 mL
1 L
Description
TraceSELECT™ Ultra and TraceSELECT
- ultra-pure acids, bases, and salts for smelting and wet digestion in environmental, water, and food analysis
- sample preparation for trace analysis requires reagents of the highest purity; our TraceSELECT Ultra products for ultra-trace analysis at ppb and even ppt levels are produced by sub-boiling distillation
- the acids, bases, and salts in the TraceSELECT series have been developed for sample preparation and analysis in the ppb (μg/kg) trace range
- packaged in high-quality containers appropriate for each product
- the reagents are produced and bottled under clean-room conditions
Specifications
Density | 1.685g/mL at 25°C |
Vapor Pressure | 2.2 mmHg (20°C), 5 mmHg (25°C) |
Quantity | 250 mL |
Linear Formula | H3PO4 |
UN Number | UN1805 |
Beilstein | 1921286 |
Formula Weight | 98g/mol |
Percent Purity | ≥85% (T) |
Grade | TraceSELECT™ Ultra |
Chemical Name or Material | Phosphoric acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'