missing translation for 'onlineSavingsMsg'
Learn More

Phosphoric Acid, N.F. - F.C.C., J.T.Baker™

Brand:  JT Baker 0262-04

Additional Details : Weight : 4.00000kg Transport : UN number : 1805 Chem class : 8 Pack group : III

Packaging Poly Bottle
Quantity 4L

 View more versions of this product

Product Code. 15577794

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Specifications

Chemical Identifiers

7664-38-2
97.994
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
CHEBI:26078
OP(=O)(O)O
H3O4P
NBIIXXVUZAFLBC-UHFFFAOYSA-N
1004
phosphoric acid

Specifications

Phosphoric Acid
7664-38-2
orthophosphoric acid, sonac, o-phosphoric acid, phosphorsaeure, acidum phosphoricum, evits, wc-reiniger, acide phosphorique, white, acido fosforico
OP(=O)(O)O
97.994
CHEBI:26078
FCC/NF
1L = 1.69kg
4L
H3O4P
NBIIXXVUZAFLBC-UHFFFAOYSA-N
phosphoric acid
1004
98
Poly Bottle
Videos
Documents