missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphoric Acid, Multi-Compendial, NF, J.T. Baker™
312.55 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
Quantity:
500 mL
2.5 L
4 L
Packaging:
Glass Bottle
Glass Bottle, Single ship
PVC Coated Glass Bottle
Poly Bottle
Specifications
Packaging | Glass Bottle |
Specific Gravity | 1L = 1.69kg |
Quantity | 2.5 L |
Formula Weight | 98 |
Grade | Multi-Compendial/NF |
Chemical Name or Material | Phosphoric Acid |
Safety and Handling
Signal Word
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'