missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phosphonoacetic acid, 98+%, Thermo Scientific Chemicals
43.33 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
10 g
25 g
100 g
Description
Phosphonoacetic acid inhibits deoxyribo nucleic acid polymerase, especially in viruses. It is used as an antiviral agent. Further, it is used in the preparation of 1-(dimethylamino)-2,2-bis[(dimethylimmonio)-methyl]ethene diperchlorate by reacting with phosphorus oxychloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phosphonoacetic acid inhibits deoxyribo nucleic acid polymerase, especially in viruses. It is used as an antiviral agent. Further, it is used in the preparation of 1-(dimethylamino)-2,2-bis[(dimethylimmonio)-methyl]ethene diperchlorate by reacting with phosphorus oxychloride.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Phosphonoacetic acid inhibits deoxyribo nucleic acid polymerase, especially in viruses. It is used as an antiviral agent. Further, it is used in the preparation of 1-(dimethylamino)-2,2-bis[(dimethylimmonio)-methyl]ethene diperchlorate by reacting with phosphorus oxychloride.
Solubility
Soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 139°C to 146°C |
Odor | Odorless |
Quantity | 10 g |
UN Number | UN3261 |
Beilstein | 1764355 |
Sensitivity | Hygroscopic |
Solubility Information | Soluble in water. |
Formula Weight | 140.04 |
Percent Purity | ≥98% |
Chemical Name or Material | Phosphonoacetic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'